17-1321-01
17-1321-01EA
97.28
EUR
InStock
17-1321-01
Tris-(hydroxyméthyl) aminométhane (TRIS, Trométamol)
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Tris (tris[hydroxymethyl] aminoethane) is suitable for preparing electrophoresis buffer in the pH range of 7,2 to 9,0. It is qualified for 2-D electrophoresis.