Multimodal chromatography columns, HiTrap™ Capto Core 400
Fournisseur: Cytiva
17-3724-11
17-3724-11EA 454 EUR InStock
17-3724-11
Multimodal chromatography columns, HiTrap™ Capto Core 400
Colonnes pour chromatographie
Novel core bead technology and multimodal ligand give Capto Core 400 dual functionality, size exclusion and binding chromatography, in one chromatography resin.
  • High capacity and productivity in flowthrough mode
  • Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification
  • Wide operational window of pH and conductivity
Capto Core 400 is an excellent alternative to size‑exclusion chromatography (SEC) resins that are typically employed in the final stages of virus purification in vaccine manufacture.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité