Anion exchange chromatography columns, SOURCE™ 15Q PE
Fournisseur: Cytiva
17-5181-01
17-5181-01EA 1543 EUR InStock
17-5181-01
Anion exchange chromatography columns, SOURCE™ 15Q PE
Colonnes pour chromatographie
SOURCE™ 15Q PE is a strong anion exchanger prepacked in a tricorn column for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.
  • Improved stability in organic solvents and pH extremes
  • Fast, high-resolution purifications and easy scale-up
  • Improved capacity compared to MonoBeads but with a slightly lower resolution
Rigid, monodispersed, spherical particles with controlled pore-size of 15 µm offers excellent flow characteristics and sample loading of up to 25 mg of protein/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité