Hydrophobic interaction chromatography columns, SOURCE™ 15PHE PE
Fournisseur: Cytiva
17-5186-01
17-5186-01EA 2116 EUR InStock
17-5186-01
Hydrophobic interaction chromatography columns, SOURCE™ 15PHE PE
Colonnes pour chromatographie
SOURCE 15PHE 4.6/100 PE is a Tricorn high performance column pre-packed with SOURCE 15PHE.
  • High resolution separations at high loads and high flow rates
  • Wide range of selectivities
  • Reproducible quality
  • Excellent scalability
  • High chemical and physical stability
  • Low backpressure
SOURCE 15PHE 4.6/100 PE high capacity makes it an excellent choice for preparative hydrophobic interaction purifications at laboratory scale. In addition, its longer bed height provides increased resolution. The column is also useful for optimization studies during scale-up.
Information de livraison: Valco fittings for ÄKTAdesign systems and are supplied with M6 connectors for FPLC system.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité