Affinity chromatography media, Capto™ Blue
Fournisseur: Cytiva
17-5448-01
17-5448-01EA 248 EUR InStock
17-5448-01
Affinity chromatography media, Capto™ Blue
Protéines purification Protein Purification Resins
Capto™ Blue is an affinity medium for the capture of albumin, purification of enzymes, and recombinant proteins at laboratory and process scales.
  • Excellent chemical stability ensures tolerance of harsh solvents used in repeated cleaning-in-place and sanitisation procedures
  • Can be autoclaved repeatedly
  • Highly rigid agarose base matrix allows high flow rates and processing of large sample volumes with no reduction in binding capacity
  • Ligand functionality may be modified through the use of appropriate buffer salts and buffer conductivity to increase selectivity for desired targets
Excellent choice for the removal or purification of proteins at both laboratory and process scales.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité