Anion exchange chromatography column, HiScreen™ Capto™ Q ImpRes
Fournisseur: Cytiva
17-5470-15
17-5470-15EA 155 EUR InStock
17-5470-15
Anion exchange chromatography column, HiScreen™ Capto™ Q ImpRes
Colonnes pour chromatographie
HiScreen™ Capto™ Q ImpRes columns are prepacked with Capto™ Q ImpRes strong anion exchange BioProcess medium. An excellent choice for method optimization and parameter screening.
  • Capto™ Q ImpRes is designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification
  • Columns with bed height of 10 cm, is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité