Multimodal columns, HiScreen™ Capto Core 700
Fournisseur: Cytiva
17-5481-15
17-5481-15EA 204 EUR InStock
17-5481-15
Multimodal columns, HiScreen™ Capto Core 700
Colonnes pour chromatographie
Capto Core 700 is designed for intermediate purification and polishing of viruses and other large biomolecules.
  • For intermediate purification and polishing of viruses and other large biomolecules (Mr>700000) in flowthrough mode
  • Significantly improved productivity and higher flow rates compared with gel filtration methods
  • Straightforward optimization due to flowthrough chromatography and robust performance
  • Convenient small-scale purification, process development and scale-up using prepacked HiTrap and HiScreen columns
The novel core bead technology and multimodal, octylamine ligand give Capto Core 700 dual functionality, size exclusion and binding chromatography in one chromatography medium (resin).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité