Set de sélection HiTrap™ IEX
Fournisseur: Cytiva
17-6002-33
17-6002-33EA 306 EUR InStock
17-6002-33
Set de sélection HiTrap™ IEX
Colonnes pour chromatographie
HiTrap IEX Selection Kit is used for screening of the optimal Cytiva ion exchange resin for specific application and development work.
  • Fast and easy selection: Seven prepacked HiTrap columns, each containing a different Sepharose ion exchange resin (medium)
  • Convenient way to screen for the optimal Cytiva anion exchange (aex) or cation exchange (cex) resin for a given application
  • Excellent for optimization studies; buffer composition, pH, flow rate, and other parameters can be optimized with small samples before scale-up
  • HiTrap column format is designed for use with a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
Cytiva's HiTrap IEX Selection Kit contains a variety of anion and cation exchange resins, allowing users to screen for the most appropriate column.
Prepacked HiTrap IEX columns are also available individually. The ion exchange resins in the columns are offered as bulk resin and in other prepacked formats.
Information de livraison: The kit includes seven 1 ml HiTrap columns packed with the following ion exchange resins: SP Sepharose Fast Flow cex column (strong cation exchanger), SP Sepharose XL cex column (strong cation exchanger), CM Sepharose Fast Flow cex column (weak cation exchanger), Q Sepharose Fast Flow aex column (strong anion exchanger), Q Sepharose XL aex column (strong anion exchanger), DEAE Sepharose Fast Flow aex column (weak anion exchanger), ANX Sepharose 4 Fast Flow aex column (weak anion exchanger).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité