17093-1L
17093-1LEA
117
EUR
InStock
17093-1L
Ammoniaque ≥25% (base NH₃) dans l'eau, Puriss. p.a. plus, Fluka™
Ammoniaque
- Exceeds standard ACS specifications with additional trace metals analysis
- Products with a guarantee certificate and/or suitable for the stated analytical application.