Cleaning solution for ultrapure water systems, Arium®
Fournisseur: Sartorius
611CDS1
171-1024EA 39.7 EUR InStock
171-1024
Cleaning solution for ultrapure water systems, Arium®
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
The Arium® cleaning solution is used to effectively remove impurities that can built up over time during water treatment.
  • Highly effective cleaning
  • Free of organic components
  • Gentle to the materials
Regular cleaning not only removes impurities but also leads to a longer service life of consumables and increases the flow rate.
Filled and ready to use in the practical cleaning syringe, the cleaning solution can be used directly.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité