Ultrapure water systems, Barnstead™ Smart2Pure™
Fournisseur: Thermo Fisher Scientific
50129688 50129845 50129885 50129887 50129890 50157872 50157873 50157877 50157882
171-1033EA 7630 EUR InStock
171-1033 171-1034 171-1028 171-1035 171-1029 171-1295 171-0993 171-0994 171-0995
Ultrapure water systems, Barnstead™ Smart2Pure™
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
The Barnstead Smart2Pure™ system is a compact system that converts tap water into both ASTM Type I and Type II water. It produces 3 or 6 liters of ultra-pure water per hour.
  • Validated 0,2 µm final filter can be autoclaved up to 5 times
  • UV-Oxidation reduces organic compounds in the water to ultra-low levels as well as microorganisms and their metabolites (on UV systems only)
  • Internal ultrafilter (UF) is flushed automatically and has a large surface area to ensure the highest retention of endotoxins and nucleases (on UF systems only)
  • Dispensing is easy and features variable speed controlled flow
  • Display can be tilted for optimal reading
  • Space-saving design: Can be positioned on the bench or mounted to the wall
Smart2Pure™ 3 and 6 systems feature a built-in 6 L reservoir to store the Type 2 water. The Smart2Pure 12 system can be combined with a 30 or 60 L reservoir for added storage capacity.
Water purification progresses through independent cartridges with Aquastop quick-connect for fast replacement:
- Module 1 offers a combination of pre-treatment and the reverse osmosis membrane
- Module 2 includes a polishing cartridge that contains high quality ultrapure resin for consistent purity and long cartridge life
Ideal for molecular biology, microbiology and analytical chemistry applications.
Information de livraison: Systems include RO/pre-treatment cartridge, an ultrapure polisher cartridge, sterile 0,2 µM filter, and UV lamp and/or UF filter where applicable. 3 and 6 L/h version also include an internal 6 L tank with vent filter.
Smart2Pure™ Pro systems are complete systems including a 30 or 60 L reservoir.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité