17132.30
17132.18
17132.30EA
275
EUR
InStock
17132.30
17132.18
Bismuth (III) oxyde ≥99.99% (base métaux)
Bismuth (III) oxyde
Purity: 99.99%
CAS No.: 1304-76-3
Molecular Formula: Bi2O3
Formula Weight: 465.96
Physical Form: -325 Mesh Powder
Density: 8.9
Melting Point: 817°
Boiling Point: ca 1890°
Merck Reference: 14,1273
MDL No.: MFCD00003462