Synonyms:
2,6-Dimethylhept-5-enal
17773.30
17773.30EA
98.9
EUR
InStock
17773.30
2,6-Diméthyl-5-heptenal stabilisé
2,6-Diméthyl-5-heptenal
CAS No.: 106-72-9
Molecular Formula: C9H16O
Formula Weight: 140.22
Flash Point: 60°(141°F)
Physical Form: Liquid
Density: 0.879
Refractive Index: 1.4441
Sensitivity: Air Sensitive
Boiling Point: 116-124°/100mm
UN No.: 1989
MDL No.: MFCD00006981