Rotary vane pump kit, E-LAB 2
Fournisseur: EDWARDS HIGH VACUUM METTLER
A65207903
181-0359EA 8740 EUR InStock
181-0359
Rotary vane pump kit, E-LAB 2
Pompe Pompes à vide à palettes
Pump includes all of the components required for laboratory applications such as those found in wet chemistry and sample preparation. It allows for continuous running even in the presence of water and solvents and incorporates an integral fast acting inlet valve that holds the system under vacuum in case of a power failure. Ideal for use with rotary evaporators, centrifugal concentrators, vacuum ovens, distillation/extraction apparatus, fume hoods, glove boxes and vacuum filtration equipment.
  • Quiet in operation, less than 50 dB
  • High efficiency oil mist filter, no oily smells in the laboratory
  • Continuous oil return kit, infrequent oil changes
  • Internal cartridge design, easy to service
W×D×H: 30×158×450 mm
Weight: 22 kg
Inlet/outlet connectors I×Ø=12,7 mm
Information de livraison: Supplied complete with outlet mist filter, oil return kit, inlet trap, oil change tag, nozzles, elbow, all fittings, one bottle of oil and instruction manual.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité