Scroll pump, XDS series
Fournisseur: EDWARDS HIGH VACUUM METTLER
A72401903
181-0362EA 0 EUR InStock
181-0362
Scroll pump, XDS series
Pompe Pompes à vide à membrane
These XDS series pumps are completely oil free, dry scroll and improved on their pumping speeds, combined with lower ultimate pressures, lower power consumption and lower noise. Gas ballast allows for pumping of condensable vapours including water, solvents, dilute acids and bases. It also feature the latest in tip seal technology giving significantly longer life between tip seal changes. Comes with integrated inverter drive with auto sensing voltage input delivers optimised pumping performance. The pumps are completely field serviceable.
  • Better work environment and low environmental impact
  • Simple operation and easy to use controls
  • Hermetically sealed for a lubricant-free vacuum environment
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité