VARIO® chemistry pumping unit PC 3001 VARIOpro
Fournisseur: VACUUBRAND
20696700 696702 696701 738401 733501 737501 20738400 733502 737502 738402 20737514
181-1646EA 5580 EUR InStock
181-1646 181-1647 181-1032 181-1086 181-1060 181-1081 181-1085 181-0196 181-0197 181-0198 VACU20737514
VARIO® chemistry pumping unit PC 3001 VARIOpro
Pompe Pompes à vide à membrane
This VARIO® chemistry pumping unit optimizes vacuum conditions automatically by precisely and continuously adjusting the diaphragm pump´s motor speed. Thanks to the motor speed control the pump operates only when vacuum is needed, saving energy and reducing maintenance.
  • Easily operated CVC 3000 vacuum controller with clear text menus and integrated venting valve
  • Automatic optimization of the vacuum level throughout the process for high process reproducibility and unattended operation
  • Compact and powerful; excellent ultimate vacuum, even with continuous condensate purge
  • Whisper quiet and ultra low vibration
  • Excellent environmental friendliness due to low power consumption and efficient solvent recovery
The integrated vacuum controller enables fully automatic evaporation at a push of a button. Low space requirements, light weight and low operational noise contribute further to this unit’s flexibility and popularity for laboratory use. The PC 3001 VARIOpro is ideal for vacuum applications with high boiling point solvents. The inlet separator, made of glass with a protective coating, prevents droplets and particles from entering the pump. The highly efficient insulated exhaust vapor condenser has a very compact design and provides near 100 % recovery of solvents.
Offers the most advanced control technology available with adaptive and automatic speed control.
Offers a high suction capacity, even with low aspiration pressures. Final vacuum as low as 7 mbar.
Information de livraison: Pumping unit completely mounted, ready for use, with manual.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité