Vacuum aspirator pump, Nalgene®
Fournisseur: Thermo Fisher Scientific
6140-0010
181-9930EA 24.8 EUR InStock
181-9930
Vacuum aspirator pump, Nalgene®
Pompe Systèmes de pompe à vide
Attach the Nalgene™ aspirator vacuum pump directly to sink faucets for vacuum draw. It can draw 11,5 L air per minute at a water flow rate of 6,5 L per minute. It operates efficiently with water pressures as low as 7,5 psig for minimum water consumption and comes with an integral check valve and tubing for insertion in discharge end.
  • Attaches to a standard faucet with ³/₈" NPT connecting thread for easy installation
  • Ultimate achievable vacuum of 724 mm Hg
  • Integral check valve that prevents loss of vacuum when water source is disrupted and prevents backflow
  • Polypropylene, with PE tubing
Informations de commande: Includes integral check valve, ³/₈" NPT connecting thread and 120 mm of tubing.
Conditionnement: Case of 24.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité