Supelco® TraceCERT® Organic Reference Standard, Metrafenone
Fournisseur: Merck
18996-50MG
18996-50MGEA 101 EUR InStock
18996-50MG
Supelco® TraceCERT® Organic Reference Standard, Metrafenone
Étalons Étalons organiques
Metrafenone CRM may also be used in the residue analysis of: 209 pesticides and persistent organic pollutants (POPs) extracted by QuEChERS-based procedure from fresh bovine and chicken liver samples using gas chromatography coupled to tandem mass spectrometry (GC-MS/MS). 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high-resolution tandem mass spectrometry. 250 pesticide residues extracted by QuEChERS method from 103 processed fruit samples using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS). 379 pesticides in human serum samples extracted by QuEChERS method using liquid chromatography-tandem mass spectrometry (LC-MS/MS). Metrafenone in nine fresh vegetable samples using matrix solid-phase dispersion (MSPD) and HPLC-UV. Metrafenone in bitter gourd and soil samples using gas chromatography combined with an electron capture detector (GC-ECD).
Certifications: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Attention: P273 - P391 - P501
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité