Supelco® TraceCERT® Organic Reference Standard, Monolinuron
Fournisseur: Merck
19000-50MG
19000-50MGEA 67.2 EUR InStock
19000-50MG
Supelco® TraceCERT® Organic Reference Standard, Monolinuron
Étalons Étalons organiques
Monolinuron CRM may also be used as given below: Determination of 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high-resolution tandem mass spectrometry. Extraction of 13 pesticides from four groundwater and three river water samples using silica-based MSU-1 mesoporous solid as a sorbent for their solid-phase extraction (SPE) followed by ultra-performance liquid chromatography-triple quadrupole-mass spectrometric (UHPLC- QqQ-MS/MS) determination. Development and validation of a UHPLC-diode array detection (DAD) method for simultaneous determination of urea and tebuthiuron herbicides in four fresh vegetable samples. Multi-residue analysis of 50 herbicides in 51 grain samples of soybean corn and wheat by ultra-performance liquid chromatography-electrospray ionisation-mass spectrometry (UPLC-ESI-MS). Extraction of 109 pesticide residues from different chemical classes from 345 tomato samples by modified QuEChERS method and their determination using liquid chromatography-tandem mass spectrometry (LC-MS/MS). Simultaneous analysis of 238 pesticides and 78 veterinary drugs in 80 bovine milk samples using ultra-fast liquid chromatography-tandem mass spectrometry (UFLC-MS/MS) and gas chromatography-tandem mass spectrometry (GC-MS/MS).
Certifications: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Attention: P260 - P264 - P270 - P273 - P301 + P312 - P314
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité