Anti-IgG Chicken Polyclonal Antibody (CF™ 350)
Fournisseur: Biotium
20332-500UL
20332-500ULEA 300 EUR InStock
20332-500UL
Anti-IgG Chicken Polyclonal Antibody (CF™ 350)
Anticorps
CF™350 is a blue fluorescent dye with excitation/emission maxima at 347/448nm. CF™350 is optimized for UV excitation, and is spectrally similar to AlexaFluor®350, AMCA, and DyLight®350. CF™350 is more water soluble, more photostable, and 50% brighter when conjugated to antibodies than the traditional blue dye AMCA. CF™350 antibody conjugates are comparable in brightness to AlexaFluor®350 dye conjugates.
CF™350 is a blue fluorescent dye with excitation/emission maxima at347/448 nm. CF™350 is optimized for UV excitation, and is spectrally similar to AlexaFluor®350, AMCA, and DyLight™350. CF™More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité