Anti-IgG Chicken Polyclonal Antibody (CF™ 543)
Fournisseur: Biotium
20335-500UL
20335-500ULEA 300 EUR InStock
20335-500UL
Anti-IgG Chicken Polyclonal Antibody (CF™ 543)
Anticorps
CF™543 antibodies are affinity-purified antibodies labeled with orange fluorescent CF™543 (Ex/Em 541/560nm), a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™543 is an orange fluorescent dye with an excitation spectrum optimally matching the 543 laser line. Antibody conjugates of the dye are much brighter than those ofMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité