Anti-IgG Chicken Polyclonal Antibody (CF™ 568)
Fournisseur: Biotium
20338-500UL
20338-500ULEA 300 EUR InStock
20338-500UL
Anti-IgG Chicken Polyclonal Antibody (CF™ 568)
Anticorps
CF™568 antibodies are affinity-purified antibodies labeled with red fluorescent CF™568, a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™568 is a red fluorescent dye optimized for the 568nm laser line. CF™568 is spectrally similar to AlexaFluor®568, ATTO 565, and Rhodamine Red. Antibody conjugates of CF™568 areMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité