Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Fournisseur: Biotium
20341-500UL
20341-500ULEA 466 EUR InStock
20341-500UL
Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Anticorps
CF™633 antibodies are affinity-purified antibodies labeled with far-red fluorescent CF™633, a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™633 is a far-red fluorescent dye with excitation/emission maxima at 630/650nm. CF™633 is spectrally similar to AlexaFluor®633, AlexaFluor®647, Cy®5, and DyLight®633. Compared to theseMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité