Anti-IgG Goat Polyclonal Antibody (CF™ 660C)
Fournisseur: Biotium
20368-500UL
20368-500ULEA 466 EUR InStock
20368-500UL
Anti-IgG Goat Polyclonal Antibody (CF™ 660C)
Anticorps
CF™660C and CF™660R antibodies are affinity-purified antibodies labeled with far-red fluorescent CF™660C or CF™660R dyes, members of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™660C is a far-red fluorescent dye with excitation/emission maxima at 667/685nm. CF™660C is several fold brighter and more photostable than the spectrallyMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité