Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Nous tenons à vous informer que notre plateforme d’e-commerce fera l’objet d’une procédure de maintenance planifiée entre 6:00 et 14:00 ET le 26 janvier 2025. Au cours de cette période, notre site ne sera pas disponible. Merci de votre compréhension.

Anti-IgG Goat Polyclonal Antibody (CF™ 790)

Fournisseur: Biotium
20379-50UL
20379-50ULEA 249 EUR InStock
20379-50UL
Anti-IgG Goat Polyclonal Antibody (CF™ 790)
Anticorps
Near-infrared CF™ dye antibodies are affinity-purified antibodies labeled with near-infrared fluorescent CF™750 or CF™770, two members of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.

CF™680 has excitation/emission maxima at 681/698nm, and CF™680R has excitation/emission maxima at 680/701; these dyes are spectrally similar to AlexaFluor®680, Cy®5.5, and DyLight®680, and IRDye®680LT. Both dyes are brighter than spectrally similar dyes; CF™680 is exceptionally bright, while CF™680R is extremely photostable. CF™750 has excitation/emission maxima at 755/777nm, and is spectrally similar to AlexaFluor®750, APC-AlexaFluor®750, Cy®7, and IRDye® 750. CF™770 has excitation/emission maxima at 770/797nm, and is spectrally similar to DyLight™800 and IRDye® 770. Near-Infrared dyes are useful for in vivo imaging, flow cytometry, multiplex Western blotting, and In-Cell Western™ assays. While other near-infrared dyes suffer from limited fluorescence brightness, poor stability and high background, Biotium's near-infrared CF™ dyes have been designed to overcome these problems to produce bright, stable antibody conjugates with high signal-to-noise ratio in immunostaining. CF™750 and CF™770 are compatible the the LI-COR Odyssey® near-IR detection system in combination with CF™680 or CF™680R antibody conjugates, or Biotium's far-red RedDot™1 nuclear stain for cell number normalization.

CF™ dyes are covered by pending U.S. and international patents. CF™ and RedDot™ are trademarks of Biotium, Inc. Alexa Fluor® is a registered trademark of Molecular Probes, Inc. Cy® is a trademark of GE Healthcare. DyLight® is a registered trademark of Pierce Biotechnology. In Cell Western™ is a trademark and IRDye® and Odyssey® are registered trademarks of LI-COR Biosciences.

Informations de commande: Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF790
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité