Labware value pack, Nalgene®
Fournisseur: Thermo Fisher Scientific
1350-0001
212-0440EA 171 EUR InStock
212-0440
Labware value pack, Nalgene®
Béchers
Labware Value Pack is a useful assortment of popular labware items used in every lab.
  • Receive 20 items with one catalog number
  • Serves as new lab starter or refresher pack
  • Sets the standard for quality and superior performance with the highest quality materials
  • Shatterproof and it provides a safer, longer equipment life at lower replacement costs than glass
Note: Remove jar closure and autoclave separately. LDPE Unitary™ wash bottles are not autoclavable. All other items in Value Pack are autoclavable.
Certifications: ASTM E1272, CFR 21, ISO 6706, Part 177.1520.
Informations de commande: Includes six straight-sided jars (500 ml/PPCO), four Unitary™ wash bottles (500 ml/LDPE), four graduated cylinders (2 to 100 ml, 500 ml, 1 L/PPCO), five Griffin low form beakers (50, 100, 250, 600, 1000 ml/PPCO), Unwire™ blue half-rack for 30 mm tubes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité