Test tube racks, Combi
Fournisseur: DELTALAB
330
212-9175EA 188 EUR InStock
212-9175
Test tube racks, Combi
Portoirs Tube Racks
Made of polypropylene, this combi rack can be autoclaved. This design consists of a unit of four independent cubes packaged in a clear support tray.
  • Excellent chemical resistance
  • Excellent temperature resistance
Each rack may be independently rotated to allow the simultaneous storage and use of multiple sizes of tubes (micro tubes to 50 ml tubes). Each side of the racks is alpha-numeric matrixed for better sample location.
The following tubes can be accommodated in each rack:
- 2×50 ml conical tubes,
- 6×15 ml conical tubes,
- 9×12 mm diameter tubes or 1,5/2,0 ml micro tubes
- 12×0,5 ml micro tubes.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité