TILT bottles, DURAN®
Fournisseur: DWK Life Sciences
218914459
215-3878EA 51.1 EUR InStock
215-3878
TILT bottles, DURAN®
Flacons Flacons pour milieux de culture
The TILT bottle, made of clear DURAN® borosilicate glass 3.3, and is ideal for cell culture media preparation.
  • Low bottle height for easy handling in biosafety cabinets
  • Space saving shape for fridge storage and water baths
  • 45° TILT position for pipetting without holding the bottle
  • 100% bigger opening for easy pipetting
  • Tilted volume indicators for convenience
  • Vacuum resistant bottle for safe filter sterilisation
Biosafety cabinets and clean hoods are essential for working with cell cultures under sterile conditions. Unfortunately, they make the handling of media bottles difficult and time-consuming for researchers. The TILT bottle has two positions: Upright for filter sterilisation or storage, and tilted at 45° for pipetting. Take a look at cell culture media from a new angle and discover a bottle system that eases handling and turns usability into security.
Information de livraison: Supplied complete with white GL 56 polypropylene screw cap.
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité