Hydra square bottle, GL 45, with 4 side necks, DURAN®
Fournisseur: DWK Life Sciences
218505453
215-4548EA 411 EUR InStock
215-4548
Hydra square bottle, GL 45, with 4 side necks, DURAN®
Flacons Flacons en verre
HPLC 1000 ml purging bottle with five GL 45 inlets.
  • Convenient – purging bottle accepts up to five HPLC GL 45 caps
  • Safer – capped bottles reduce the risk of volatile organic compound (VOC) escape and spillage
  • Compatible - GL 45 threads fit standard HPLC connection caps
  • Manufactured from the material of choice for HPLC solvent bottles: Borosilicate 3.3 glass offering a very low inorganic ion extractables profile
  • 1 L square space saving shape
  • Durable white enamel volume graduations (100 to 800 ml, in 100 ml intervals)
DURAN® bottles are ideal for the storage and delivery of high-performance liquid chromatography (HPLC) mobile phases and solvents. Increasing the number of GL 45 inlets on the bottles adds convenience: The five GL 45 necks on this purging bottle increases safety and reduces the risk of cross contamination and air intake.
Applications: HPLC purge bottle, reagent supply to liquid handling systems and analysers.
Certifications: Clear DURAN® is a borosilicate 3.3 glass as specified by ISO 3585, and is classified as a 'Type I, Class A low expansion borosilicate glass' as defined by ASTM-E438-1992. Retrace Code, and certificate for manufacturing lot traceability.
Glass Type I according to the requirements of the current United States Pharmacopoeia USP <660> Containers – Glass.
Glass Type I according to the requirements of the current European Pharmacopoeia EP (3.2.1) Glass Containers for Pharmaceutical Use.
Glass Type I according to the requirements of the current Japanese Pharmacopoeia JP <7.01> Test for Glass Containers for Injections.
The calibration results of volumetric instruments with this certificate refer to one calibration batch.
This batch certificate confirms that ASTM Class A error limits required by USP are met. The certificate records the mean value, standard deviation of the batch, and the date of issue.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité