VWR® 24-Hour Urine Container
Fournisseur: VWR Collection
216-2327EA 183 EUR InStock
216-2327
VWR® 24-Hour Urine Container
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons d'urine
24-hour 3 L vacuum urine container with aspiration system for vacuum tubes directly integrated in the screw cap.
  • Optimal handling: Ergonomic moulded handle
  • Graduated
  • Polyethylene
The 24 hour urine collection container is suitable for collecting urine over a timed period.
The 3 L container is brown coloured to prevent the premature decomposition of light sensitive parameters such as urobilinogen.
The container is fitted with an integrated transfer device to enable hygienic, clean sample transfer into a urine tube.
Efficiency in collecting biological samples.
New translucent vertical stripe: This solution allows operators to control the level of urine inside the container, avoiding the risk of contamination, speeding up operations and workflow in the laboratory.
Certifications: IVD
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité