Catalase (from Aspergillus niger)
Fournisseur: Merck Millipore Calbiochem
219261-100
219261-100EA 170 EUR InStock
219261-100
Catalase (from Aspergillus niger)
Enzymes
Catalase, fungal suspension from from Aspergillus niger is long-acting, extremely stable form of catalase that is active over a wide pH range (pH 1 to 12). Composed of four protein subunits, each containing a heme group bound to its active site.
Catalase is used for the removal of peroxides, the generation of oxygen and, in coupled systems, for the determination of metabolites e.g. uric acid. A very stable preparation particularly suitable for immobilisation.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité