Synonyms:
Cadmium arsenide
22773.09
22773.09EA
184
EUR
InStock
22773.09
Arséniure de cadmium (II) ≥99% (base métaux)
Arséniure de cadmium (II)
Purity: 99%
CAS No.: 12006-15-4
Molecular Formula: As2Cd3
Formula Weight: 487.04
Physical Form: Lump
Density: 3.031
Appearance: Dark grey
Odour: Odourless
UN No.: 1557
MDL No.: MFCD00049723