22920.K2
22920.K2EA
83.1
EUR
InStock
22920.K2
Chloroforme ≥99.5% pour HPLC
Chloroforme
Purity: 99.5+% min
CAS No.: 67-66-3
Molecular Formula: CHCl3
Formula Weight: 119.38
Physical Form: Liquid
Density: 1.492
Refractive Index: 1.4460
Odour: Characteristic
Melting Point: -63°
Boiling Point: 60.5-61.5°
Beilstein Reference: 1731042
Merck Reference: 14,2141
UN No.: 1888
MDL No.: MFCD00000826