22955.30
22955.30EA
314
EUR
InStock
22955.30
Tungsten carbide cobalt ≥99% (base métaux)
Tungsten carbide cobalt
Purity: 99%
CAS No.: 12774-15-1
Molecular Formula: WC:Co; 94:6 wt%
Physical Form: -325 Mesh Powder
Appearance: Dark grey
Odour: Odourless
UN No.: UN3178
MDL No.: MFCD00064847