Kit de spatules
Fournisseur: RSG
000.105
231-2176EA 48.9 EUR InStock
231-2176
Kit de spatules
Spatules
Stainless steel 18/10. Useful combination of double spatulas, spoon spatulas and forceps. Universal usage in laboratories and industry.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Comes in practical imitation leather case
Each set includes a 150×5 and 150×7 mm spoon spatula, a 150×9 mm double spatula, a 150×4 mm micro double spatula, a 150×9 mm powder spatula, a 150×4 mm micro powder spatula, as well as tweezers (straight, blunt, 145 mm long).
Each set includes a 150×5 and 150×7 mm spoon spatula, a 150×9 mm double spatula, a 150×4 mm micro double spatula, a 150×9 mm powder spatula, a 150×4 mm micro powder spatula, as well as forceps (straight, blunt, 145 mm long).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité