Multi-purpose scissors, Rubis®
Fournisseur: OUTILS RUBIS SA
1C200/EL
233-0218EA 39 EUR InStock
233-0218
Multi-purpose scissors, Rubis®
Ciseaux
Long legged multi-purpose scissors with rounded-off tips.
  • Produced from hardened stainless steel AISI420
  • Highly resistant against acids and alkaline
  • Ergonomically designed Rubis Twist® grips
  • No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under a magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité