Scissors with tweezer tips, Rubis®
Fournisseur: OUTILS RUBIS SA
1K603
233-0226EA 28.9 EUR InStock
233-0226
Scissors with tweezer tips, Rubis®
Ciseaux
Allround tweezers engineered into a scissors-movement.
  • Produced from hardened stainless steel AISI420
  • Highly resistant against acids and alkaline
  • With ergonomically designed Rubis Twist® grips and high precison tweezers tips
  • No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under the magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité