Vydac 238MS C18, HPLC columns
Fournisseur: Avantor
238MS5.310EA 1110 EUR InStock
238MS5.310 238GK54MS/N
Vydac 238MS C18, HPLC columns
Colonnes pour chromatographie
These Vydac MS columns are a later development from Grace for the reversed phase HPLC separation of biomolecules. A proprietary surface treatment and bonding process gives Vydac MS columns a unique selectivity. A variety of reversed phases makes this product line suitable for the analysis of small peptides to large intact, undenatured proteins.
  • Five reversed-phase chemistries
  • Excellent peak shape with little or no TFA
  • High protein recoveries make scale-up easy
Monomeric bonding offers increased peptide interaction and generally yields higher peak counts. Use for same applications as 218MS, but offers different C18 selectivity.
Vydac is a registered trademark of W.R.Grace and Co.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité