RNA isolation, illustra™ RNAspin 96 Kit
Fournisseur: Cytiva
25-0500-75
25-0500-75EA 1491 EUR InStock
25-0500-75
RNA isolation, illustra™ RNAspin 96 Kit
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
illustra™ RNAspin 96 kit is a flexible, high-throughput solution for total RNA isolation.
  • Fast and efficient purification in a 96-well format
  • 96 samples can be parallel-processed with high reproducibility in under 70 minutes
  • Flexible processing by vacuum or centrifugation
  • Integrated wash plate eliminates risk of contamination
  • DNase I treatment removes genomic DNA, leading to high quality total RNA product
illustra™ RNAspin 96 isolation kit can be used to reproducibly isolate total RNA from cultured cells, tissue, bacteria or yeast. The isolated RNA is of a high enough quality and quantity (up to 100 μg) for multiple downstream experiments, including sensitive enzymatic applications like quantitative reverse transcription polymerase chain reaction (qRT-PCR), primer extension, and RNase protection assays.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité