251402-1L
251402-1LEA
126
EUR
InStock
251402-1L
Trichloroéthylène ≥99.5% ACS, Riedel-de Haën™
Trichloroéthylène
ACS GENERAL USE SOLVENTS
- Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- Suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested to meet ACS general use specifications