Filter plates, 96-well, Protein G HP MultiTrap™
Fournisseur: Cytiva
28-9031-35
28-9031-35EA 2453 EUR InStock
28-9031-35
Filter plates, 96-well, Protein G HP MultiTrap™
Microplaques Plaques de filtration
Protein G Sepharose™ HP has high affinity for the Fc region of IgG from a variety of species.
  • Fast, reliable ease of use format in prepacked 96-well filter plates.
  • Flexible format, protocol for classical elution as well as cross-linking method
  • Formatted for elution for electrophoresis or LC-MS analysis workflows
  • Fast binding kinetics and high capacity provides high yield
  • Easy scale-up with HiTrap™ Protein G HP prepacked columns
  • Prepacked with Protein G Sepharose High Performance for coupling of antibodies of IgG subclasses
  • MultiTrap™ 96-well filter plates
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité