Kit de purification illustra™ PCR GFX™ 96
Fournisseur: Cytiva
28-9034-45
28-9034-45EA 1944 EUR InStock
28-9034-45
Kit de purification illustra™ PCR GFX™ 96
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
The illustra™ GFX™ 96 Purification Kit utilises glass fibre matrix technology in a 96-well format for highly efficient purification of PCR products. Fragments from PCR are captured by the matrix in the presence of a chaotropic salt, and contaminants are conveniently removed by washing the matrix with a buffered ethanol solution.
  • For the purification of up to 96 PCR products (0,1 to 10 kb) simultaneously in as little as 15 minutes
  • High yields of pure DNA recovered in a small volume of water or a low-ionic strength buffer
  • Typical recoveries are >85% for PCR products 100 bp to 10 kb in length; salt removal typically >99%
  • Avoids ethanol precipitations and hazardous organic extractions
Purified DNA is ready for use in most applications, including fluorescent sequencing, microarrays, labelling, hybridisation, ligation, and transformation.
Conditionnement: Kit contains sufficient quantities of binding plates (10), wash plates (10), collection plates (10), capture buffer (blue, 288 ml), wash buffer (yellow, 80 ml), a choice of two elution buffers each 48 ml (Tris-HCl and sterile water) and an instruction booklet.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité