Protein A column, HiScreen™ MabSelect SuRe™
Fournisseur: Cytiva
28-9269-77
28-9269-77EA 1322 EUR InStock
28-9269-77
Protein A column, HiScreen™ MabSelect SuRe™
Colonnes pour chromatographie
The HiScreen MabSelect SuRe column uses protein A chromatography for antibody purification. This protein A column is prepacked with MabSelect SuRe affinity resin and is part of our process development platform. These packed columns are used for method optimization and parameter screening for capture of monoclonal antibodies.
  • The novel, alkali-tolerant recombinant protein A ligand of MabSelect Sure allows cleaning-in-place (CIP)
  • Prepacked, ready-to-use columns for convenient process development
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography resin using the same linear fluid velocity
The small column volume of 4,7 ml and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up of the protein purification process. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité