Anion exchange chromatography column, HiScreen™ Capto™ DEAE
Fournisseur: Cytiva
28-9269-82
28-9269-82EA 156 EUR InStock
28-9269-82
Anion exchange chromatography column, HiScreen™ Capto™ DEAE
Colonnes pour chromatographie
HiScreen™ Capto™ DEAE column is a weak anion exchanger with a bed height of 10 cm making it an excellent tool for method optimisation, parameter screening, robustness testing, and convenient scale-up.
  • Prepacked, ready-to-use columns for convenient process development optimisation
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Easily connected in series to achieve 20 cm bed height
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity
  • Used in an optimal way with liquid chromatography systems such as ÄKTA™
  • Flow velocity: <700 cm/h
Process fluid velocities can be applied, since the bed height gives enough residence time and the results can then serve as basis for linear process scale-up. It is prepacked with Capto™ DEAE ion exchange chromatography medium.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité