Ion exchange chromatography column selection kit, HiTrap™ Capto™
Fournisseur: Cytiva
28-9343-88
28-9343-88EA 300 EUR InStock
28-9343-88
Ion exchange chromatography column selection kit, HiTrap™ Capto™
Colonnes pour chromatographie
HiTrap Capto IEX Selection Kit is designed for screening the optimal Capto ion exchange or Cytiva multimodal chromatography resin prior to scale-up.
  • Fast and easy selection. Five prepacked HiTrap columns, each containing a different IEX resin, including multimodal ion exchangers
  • Convenient way to screen for the optimal Cytiva IEX resin for a given application
  • Excellent for optimization studies, buffer composition, pH, flow rate, and other parameters can be optimized with small samples before scale-up
  • HiTrap column format is designed for use with a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
This kit contains a variety of columns prepacked with anion and cation exchange resins, allowing users to screen for the most appropriate column prior to scale-up.
Information de livraison: Kit includes five 1 ml HiTrap™ columns prepacked with Capto™ Q (strong anion), Capto™ S (strong cation), Capto™ DEAE (weak anion), Capto™ MMC (multimodal cation), Capto™ adhere (multimodal strong anion), as well as connectors and instructions.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité