Anion exchange chromatography column, HiPrep™ IEX Q Sepharose™ XL
Fournisseur: Cytiva
28-9365-38
28-9365-38EA 723 EUR InStock
28-9365-38
Anion exchange chromatography column, HiPrep™ IEX Q Sepharose™ XL
Colonnes pour chromatographie
HiPrep™ Q XL 16/10 is a prepacked, ready to use column for ion exchange chromatography.
  • Provide high loading capacity and fast purifications
  • Optimised for reliable, reproducible separations
  • Compatible with simple pump-based configurations and chromatography systems such as ÄKTA design
  • Excellent choice for rapid enrichment during initial capture of proteins from start material
  • Strong quaternary ammonium (Q) anion exchanger
It is prepacked with Q Sepharose™ XL for initial capture of proteins and other biomoleculs with ion exchange (IEX) chromatography.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité