Cation exchange chromatography column, HiPrep™ SP FF
Fournisseur: Cytiva
28-9365-44
28-9365-44EA 733 EUR InStock
28-9365-44
Cation exchange chromatography column, HiPrep™ SP FF
Colonnes pour chromatographie
HiPrep™ SP FF are ready to use strong cation exchange columns prepacked with SP Sepharose™ Fast Flow for fast, preparative separations of proteins and other biomolecules using ion exchange (IEX) chromatography.
  • HiPrep™ columns provide fast, reliable and reproducible preparative ion exchange separations
  • Good flow rates and high capacity
  • The industry standard for ion exchange chromatography
Compatible with single-pump configurations and chromatography systems such as ÄKTA design. Use a weak ion exchanger if the selectivity of a strong ion exchanger isMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité