Hydrophobic interaction chromatography columns, HiPrep™ Butyl FF
Fournisseur: Cytiva
28-9365-47
28-9365-47EA 737 EUR InStock
28-9365-47
Hydrophobic interaction chromatography columns, HiPrep™ Butyl FF
Colonnes pour chromatographie
HiPrep™ Butyl FF (High Sub) 16/10, are prepacked Butyl Sepharose™ 4 Fast Flow columns ready to use for hydrophobic interaction chromatography (HIC).
  • 20 ml HIC columns provide fast, preparative separation of proteins and other biomolecules
  • Provide high capacity, excellent flow properties, and reliable, reproducible separations
  • Butyl Sepharose™ 4 Fast Flow is a well-established, standard aliphatic HIC medium for capture and intermediate purification
  • Compatible with simple pump-based configurations or chromatography system such as ÄKTA design
The columns are designed for initial and intermediate purification, removing bulk impurities.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité