Hydrophobic interaction chromatography columns, HiPrep™ Octyl FF
Fournisseur: Cytiva
28-9365-48
28-9365-48EA 737 EUR InStock
28-9365-48
Hydrophobic interaction chromatography columns, HiPrep™ Octyl FF
Colonnes pour chromatographie
HiPrep™ Octyl FF (High Sub) 16/10, are prepacked Octyl Sepharose™ 4 Fast Flow columns ready to use for hydrophobic interaction chromatography (HIC).
  • 20 ml HIC columns provide fast, preparative separation of proteins and other biomolecules
  • Provide high capacity, excellent flow properties, and reliable, reproducible separations
  • Octyl Sepharose™ 4 Fast Flow is a well-established, standard aliphatic HIC medium for capture and intermediate purification
  • Compatible with simple pump-based configurations or chromatography system such as ÄKTA design
  • The octyl derivative adds a different and complementary selectivity to the Sepharose™ Fast Flow HIC product family
The columns are designed for initial and intermediate purification, removing bulk impurities.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité