Affinity chromatography column, HiScreen™ IMAC FF
Fournisseur: Cytiva
28-9505-17
28-9505-17EA 186 EUR InStock
28-9505-17
Affinity chromatography column, HiScreen™ IMAC FF
Colonnes pour chromatographie
HiScreen™ IMAC FF columns for optimising purification of histidine-tagged proteins by Immobilised Metal ion Affinity Chromatography (IMAC).
  • Conveniently charge with metal ion of choice (Cu²⁺, Zn²⁺, Ni²⁺ or Co²⁺)
  • Prepacked HiScreen™ columns for method optimization and process development
  • High binding capacity
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity
  • Flow velocity: <600 cm/h; The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used
The columns are prepacked with IMAC Sepharose™ 6 Fast Flow.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité